Learn More
Bioworld Nylon Membrane Stripping Buffer (10X), 250 mL
Catalog No. 50196837
Encompass_Preferred
Product DescriptionNylon Membrane Stripping Buffer (10X) is a liquid used for the hybridization of probes to nucleic acids. Buffer contains Tris HCL at pH 8.0 (1mM), EDTA (100mM), and Denhardt’s solution (1X). To use this product, dilute at 10-fold with sterile deionized water and strip the membrane by 2 hours of incubation at 75°C.
Catalog No. 50-196-837 Supplier Bioworld Supplier No. 107500161
Product DescriptionNylon Membrane Stripping Buffer (10X) is a liquid used for the hybridization of probes to nucleic acids. Buffer contains Tris HCL at pH 8.0 (1mM), EDTA (100mM), and Denhardt’s solution (1X). To use this product, dilute at 10-fold with sterile deionized water and strip the membrane by 2 hours of incubation at 75°C.