Learn More
Sigma Aldrich Fine Chemicals Biosciences Nylon Membranes for Colony
Catalog No. 502901140
Encompass
Nylon membrane is a circular uncharged microporous amphoteric ultra-pure nylon 66 membrane on a polyester support. It has a durable structure with consistent transfer rate.Surface Properties Pore size of 1.2¼m. Surface contains 50% amino and 50% carboxyl groups.
Catalog No. 50-290-1140 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11699075001
Nylon membrane is a circular uncharged microporous amphoteric ultra-pure nylon 66 membrane on a polyester support. It has a durable structure with consistent transfer rate.Surface Properties Pore size of 1.2¼m. Surface contains 50% amino and 50% carboxyl groups.