Learn More
Sigma Aldrich Fine Chemicals Biosciences Nylon Membranes positively
Catalog No. 502914536
Encompass
Microporous nylon 66 membrane on a polyester support carrying positively charged (pH 3 - 10) Aquaternary ammonium groups.Surface Properties Pore size of A0.45mum
Catalog No. 50-291-4536 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11417240001
Microporous nylon 66 membrane on a polyester support carrying positively charged (pH 3 - 10) Aquaternary ammonium groups.Surface Properties Pore size of A0.45mum