Learn More
Sigma Aldrich Fine Chemicals Biosciences Nylon Membranes positively
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11209272001
Encompass
Microporous nylon 66 membrane on a polyester support carrying positively charged (pH 3 - 10) Aquaternary ammonium groups.Surface Properties Pore size of A0.45mum
Catalog No. 50-290-5792