Learn More
ELKO FILTERING CO NYLON MESH 150 MICRON 1-4 YARD
SDP
Supplier:  ELKO FILTERING CO 03150/50
Encompass
Nylon Mesh 150 Micron - Open Area %: 50 - Width: 40 in (1-4 Yards)Price is per Yard.
Catalog No. NC1280170