Learn More
ELKO FILTERING CO NYLON MESH 20 MICRON PER YARD
SDP
Supplier:  ELKO FILTERING CO 0320/14
Encompass
Nylon Monofilament Precision Woven Mesh - 20 Micron Opening, Open Area 14%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-20/14
Catalog No. NC0569344