Learn More
ELKO FILTERING CO NYLON MESH 200UM 54OPEN 1YARD
SDP
Catalog No. NC0567492
Encompass
Nylon Monofilament Precision Woven Mesh - 200 Micron Opening, Open Area 54%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-200/54
Catalog No. NC0567492 Supplier ELKO FILTERING CO Supplier No. 03200/54
Nylon Monofilament Precision Woven Mesh - 200 Micron Opening, Open Area 54%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-200/54