Learn More
ELKO FILTERING CO NYLON MESH 300 MICRON
SDP
Supplier:  ELKO FILTERING CO 06300/34
Encompass
Nylon Monofilament Precision Woven Mesh - 300 Micron Opening, Open Area 34%, 45 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 06-300/34
Catalog No. NC0707038