Learn More
ELKO FILTERING CO NYLON MESH 37 MICRON 1 YRD
SDP
Supplier:  ELKO FILTERING CO 0337/24
Encompass
Nylon Monofilament Precision Woven Mesh - 37 Micron Opening, Open Area 24%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-37/24
Catalog No. NC0515677