Learn More
Component Supply Nylon Mesh, 40µ 26%
SDP
Supplier:  Component Supply UCMN40
Encompass
Mesh opening: 40µ; Open area: 26%; Mesh Count: 300; Thread Diameter: 38µ; Monofilament, woven mesh material is nylon 6/6 and possesses effective abrasion and alkali resistance. The softening point is between 170°C and 230°C and the melting point is between 210°C and 250°C.
Catalog No. NC1699341