Learn More
ELKO FILTERING CO NYLON MESH 41 MICRON 1YD
SDP
Catalog No. NC0449232
Encompass
Nylon Monofilament Precision Woven Mesh - 41 Micron Opening, Open Area 31%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-41/31
Catalog No. NC0449232 Supplier ELKO FILTERING CO Supplier No. 0341/31
Nylon Monofilament Precision Woven Mesh - 41 Micron Opening, Open Area 31%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-41/31