Learn More
Gilson Company Inc No. 325 Cut-To-Order Nylon Cloth
Supplier:  Gilson Company Inc NMD #325
Encompass_Preferred
45 um Nylon Screen Cloth is an option when metallic wire cloth cannot be used for particle sizing. With its high tensile strength and superior abrasion resistance, this cloth is tough against high pH conditions but tends to stretch when wet.
Catalog No. NC0856945