Learn More
McMaster-Carr Nylon Plastic Mesh, 58 x 58 Mesh Size, 40" Wide
Catalog No. NC1744840
Encompass
Made from nylon, this mesh is wear resistant. It has openings that meet an exact size tolerance, providing accurate, consistent results in filtering and sifting applications.
Catalog No. NC1744840 Supplier McMaster-Carr Supplier No. 9318T16
Made from nylon, this mesh is wear resistant. It has openings that meet an exact size tolerance, providing accurate, consistent results in filtering and sifting applications.