Learn More
ELKO FILTERING CO Nylon Mesh 60 Micron, 1 yard
SDP
Supplier:  ELKO FILTERING CO 0360/42
Encompass
Open Area %: 42, Width: 40 in, Resistant to common solvents and to alkalies but are attacked by strong mineral acids. Good abrasion resistance and low coefficient of friction. Working temperature is typically around 239F (115C). Exposure to sunlight can cause discoloration (yellowing). Water absorption is very high. Sterilizing by autoclaving, ETO sterilizing and gamma sterilizing (Under 2.5 Megarads) can be performed.
Catalog No. NC1588564