Learn More
ELKO FILTERING CO NYLON MESH 74 MICRON PER/YARD
SDP
Supplier:  ELKO FILTERING CO 0374/21
Encompass
Nylon Monofilament Precision Woven Mesh - 74 Micron Opening, Open Area 21%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-74/21
Catalog No. NC0366804