Learn More
ELKO FILTERING CO NYLON MESH FILTER 200UM 1 YARD
SDP
Supplier:  ELKO FILTERING CO 03200/45
Encompass
Nylon Monofilament Precision Woven Mesh - 200 Micron Opening, Open Area 45%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-200/45
Catalog No. NC0148096