Learn More
Braintree Scientific Nylon Monofilament, Non-absorbable Suture, 3-0 100 yds
SDP
Catalog No. 501955982
Encompass
Monofilament Nylon Suture Nylon Monofilament, Non-absorbable Suture. Deknatel's reliable nylon suture is known for its uniform diameter, flexibility, and outstanding knot security. Material: Nylon type 6,6. Coating: None. Characteristics: Nylon exhibits uniform diameter of suture, minimal memory, and outstanding and consistent knot security. Please note: Nylon suture should not be used where permanent retention of tensile strength is required. Indications: Nylon suture is indicated for use in general soft tissue approximation and/or ligation, including use in cardiovascular, neurological, and ophthalmic procedures. Sizes: USP 2 through 8-0. Construction: Monofilament. Configuration: Non-sterile spool. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5982 Supplier Braintree Scientific Supplier No. SUTM110
Monofilament Nylon Suture Nylon Monofilament, Non-absorbable Suture. Deknatel's reliable nylon suture is known for its uniform diameter, flexibility, and outstanding knot security. Material: Nylon type 6,6. Coating: None. Characteristics: Nylon exhibits uniform diameter of suture, minimal memory, and outstanding and consistent knot security. Please note: Nylon suture should not be used where permanent retention of tensile strength is required. Indications: Nylon suture is indicated for use in general soft tissue approximation and/or ligation, including use in cardiovascular, neurological, and ophthalmic procedures. Sizes: USP 2 through 8-0. Construction: Monofilament. Configuration: Non-sterile spool. **Sold for research purposes only and not for human use**