Learn More
Electron Microscopy Sciences Nylon Polishing Cloth, PSA, 2.5" centered hole, 8"
SDP
Catalog No. 50997221
Encompass_Preferred
These polishing cloths are made of a durable napless nylon weave and provide an excellent finish on most materials. The cloth is 8" in diameter.
Type: 2.5" centered hole
Diameter: 8"
Catalog No. 50-997-221 Supplier Electron Microscopy Sciences Supplier No. 5031415
These polishing cloths are made of a durable napless nylon weave and provide an excellent finish on most materials. The cloth is 8" in diameter.
Type: 2.5" centered hole
Diameter: 8"