Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Nylon Rope Trick
GREENER_CHOICESCI_ED

Catalog No. S76784A Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S76784A 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including n-Hexane, which is known to the State of California to cause and reproductive harm.
Catalog No. S76784A Supplier Innovating Science™ Supplier No. IS7010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including n-Hexane, which is known to the State of California to cause and reproductive harm.

Includes

Two ready-to-use solutions

Requires

250 mL beaker, forceps and stirring rod

Demonstrate polymerization at a liquid-liquid interface.

  • Observe the synthesis of Nylon 610 at the interface of two immiscible liquids
  • Rope can be wound around a stirring rod as the reactants continue to polymerize
  • Total demonstration time: 20 minutes
  • Enough materials for five demonstrations

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Demonstrates polymerization at a liquid-liquid interface
Includes 1 x 25 mL solution a (1,6-hexanediamine, sodium hydroxide, water) un1760 1 x 25 mL solution b (sebacoyl chloride, hexane) un1208
Quantity 1
Time Range 20 min.
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 1 Class
Product Type Nylon Rope Trick Chemistry Kit

WARNING: This product can expose you to chemicals including n-Hexane, which is known to the State of California to cause and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.