Learn More
SPS Medical 4"x 10" Nylon Self-Seal Pouch, 100/bx
Catalog No. NC1400566
Encompass_Preferred
Manufactured for use with dry heat processes up to 420°F. Material is clear, strong and puncture resistant with self-seal adheasive closure. Complimentary external chemical indicator labels are included with every box. Pouches are notched for ease of opening after processing
This item is not returnable. View return policy
Catalog No. NC1400566 Supplier SPS Medical Supplier No. NSP410
Manufactured for use with dry heat processes up to 420°F. Material is clear, strong and puncture resistant with self-seal adheasive closure. Complimentary external chemical indicator labels are included with every box. Pouches are notched for ease of opening after processing