Learn More
SPS Medical Sterilization Pouch Dry Heat 7 x 10 1/2'' Transparent Self Seal Nylon, 100/pk
Catalog No. NC1400567
Encompass_Preferred
Nylon packaging products are manufactured for use with dry heat processes up to 420°F. Nylon material is clear, strong and puncture resistant, maintaining a durable barrier for instrumentation until point of use. Self-seal nylon pouches feature built-in adhesive tape which eliminates the need for a heat sealer or indicator tape. Each pouch is notched for ease of opening after processing, and complimentary external chemical indicator labels are included.
This item is not returnable. View return policy
Catalog No. NC1400567 Supplier SPS Medical Supplier No. NSP420
Nylon packaging products are manufactured for use with dry heat processes up to 420°F. Nylon material is clear, strong and puncture resistant, maintaining a durable barrier for instrumentation until point of use. Self-seal nylon pouches feature built-in adhesive tape which eliminates the need for a heat sealer or indicator tape. Each pouch is notched for ease of opening after processing, and complimentary external chemical indicator labels are included.