Learn More
Gilson Company Inc Nylon Sieve Cleaning Brush
Catalog No. NC1830319
Encompass
Nylon Sieve Cleaning Brush is a hardwood block brush with nylon bristles and 3.75in (95mm) straight wood handle best for cleaning No. 120 mesh and finer sieves. It is 1.875in (47.6mm) in diameter and 0.875in (22.2mm) thick, with an overall brush length of 5.9in (149mm).
Catalog No. NC1830319 Supplier Gilson Company Inc Supplier No. TSA179
Nylon Sieve Cleaning Brush is a hardwood block brush with nylon bristles and 3.75in (95mm) straight wood handle best for cleaning No. 120 mesh and finer sieves. It is 1.875in (47.6mm) in diameter and 0.875in (22.2mm) thick, with an overall brush length of 5.9in (149mm).