Learn More
MSC Cwc Solid Braid Nylon Rope, 1/8 in., White, 50 ft.
Catalog No. 5000513478
Encompass
Solid braid nylon cord is a strong, lightweight, and soft general-purpose utility rope designed for applications requiring high strength and elasticity. Its lock-stitch braided construction ensures the rope remains round, resists fraying, and performs efficiently in pulley systems. This nylon cord is engineered to withstand significant strain while providing a high degree of shock-absorbing capability.
  • Solid-braided, lock-stitch construction
  • Excellent shock-absorbing elasticity
  • Flexible and easy to tie
  • Resistant to rot, mildew, abrasions, and most chemicals
  • High strength with superior elongation compared to polyester
  • Performs smoothly over pulleys and surfaces
Catalog No. 50-005-13478 Supplier MSC Supplier No. 17233685
Solid braid nylon cord is a strong, lightweight, and soft general-purpose utility rope designed for applications requiring high strength and elasticity. Its lock-stitch braided construction ensures the rope remains round, resists fraying, and performs efficiently in pulley systems. This nylon cord is engineered to withstand significant strain while providing a high degree of shock-absorbing capability.
  • Solid-braided, lock-stitch construction
  • Excellent shock-absorbing elasticity
  • Flexible and easy to tie
  • Resistant to rot, mildew, abrasions, and most chemicals
  • High strength with superior elongation compared to polyester
  • Performs smoothly over pulleys and surfaces