Learn More
Med Vet International Nylon Sponge Forceps, 8", J0579S, Each
Catalog No. 5000584288
Encompass_Preferred
NOTE: Not intended for everyday surgical use.These instruments are useful for a variety of clinical uses at a very affordable price. These instruments will come in handy for a lot of non surgical procedures.AutoclavableWill not rustVery inexpensiveNon-repairable
Catalog No. 50-005-84288 Supplier Med Vet International Supplier No. JOR579S
NOTE: Not intended for everyday surgical use.These instruments are useful for a variety of clinical uses at a very affordable price. These instruments will come in handy for a lot of non surgical procedures.AutoclavableWill not rustVery inexpensiveNon-repairable