Learn More
IW Tremont Co Inc LabExact Nonsterile Nylon Syringe Filters, 0.22um, 25mm, 100 pack
SDP
Supplier:  IW Tremont Co Inc LEIWTES10002
Encompass_Preferred
LabExact Disposable non-sterile Nylon/Polyamide filtration media syringe filters feature Luer-Lok and pressure fitting. They are ultrasonically bonded for reliability and the filter media type is clearly inscribed on barrel. Excellent unit-to-unit and lot-to-lot reproducibility and consistency. The unique concentric funnel shaped barrel increases volume and decreases back pressure. Hydrophilic material recommended for filtration of aqueous or organic/aqueous medium polar liquids. Commonly used for general laboratory filtration and filtration of HPLC and GC samples prior to injection. Suitable for filtration of aqueous and organic-aqueous polar medium liquids and dissolution sample analysis. Suitable for use with high pH samples. Since it binds proteins it is NOT suitable for protein recovery applications.
Catalog No. 50-223-7716