Learn More
Tisch Scientific Nylon Syringe Filters, 0.22 um, 20mm, Threaded Double Luer Lock, ABS Housing, Nonsterile, 100 per pack, SPEC18126
SDP
Supplier:  Tisch Scientific SPEC18126
Encompass
Nylon Syringe Filters, 0.22um, 20mm, Threaded Double Luer Lock, ABS Housing, Nonsterile, 100 per pack
Catalog No. NC1815054