Learn More
Accuflow Systems Reinforced nylon sanitary 1.5" Bio-Clamps 10 pack
Catalog No. 501108229
Encompass_Preferred
Reinforced nylon sanitary 1.5" Bio-Clamps, designed for use in GMP Manufacturing and laboratories. These clamps can be autoclavable to 275F as well as gama irradiated. Meet USP Class VI standards. Designed for Sanitary Tri clamp applications. Pack of 10
Catalog No. 50-110-8229 Supplier Accuflow Systems Supplier No. NG150WHT
Reinforced nylon sanitary 1.5" Bio-Clamps, designed for use in GMP Manufacturing and laboratories. These clamps can be autoclavable to 275F as well as gama irradiated. Meet USP Class VI standards. Designed for Sanitary Tri clamp applications. Pack of 10