Learn More
Grainger Nylon Tread Wheel 6 720 lb.
Catalog No. 502766435
Encompass_Preferred
The Grainger 6" Nylon Tread Wheel is a heavy-duty wheel designed for material handling applications. It features a nylon tread that provides quiet operation and good traction. The wheel has a load capacity of 250 lbs and measures 6" in diameter with a 1" width face. It has a precision ball bearing and a 3/4" straight axle hole. The wheel is ideal for use on carts, workbenches, and other equipment that requires smooth-rolling heavy-duty wheels. It provides an economical solution for industrial material handling needs.
Catalog No. 50-276-6435 Supplier Grainger Supplier No. 16V305
The Grainger 6" Nylon Tread Wheel is a heavy-duty wheel designed for material handling applications. It features a nylon tread that provides quiet operation and good traction. The wheel has a load capacity of 250 lbs and measures 6" in diameter with a 1" width face. It has a precision ball bearing and a 3/4" straight axle hole. The wheel is ideal for use on carts, workbenches, and other equipment that requires smooth-rolling heavy-duty wheels. It provides an economical solution for industrial material handling needs.