Learn More
MATTERHACKERS INC NYLONX CARBON FIBR FILA 1.75MM
SDP
Catalog No. NC2599241
Encompass
NylonX is a high-strength/ engineering-grade nylon filament that is blended with chopped carbon fibers to create a durable/ shatter-resistant filament that is perfect for tough/ functional 3D printed parts.
Catalog No. NC2599241 Supplier MATTERHACKERS INC Supplier No. MPUPDPEN
NylonX is a high-strength/ engineering-grade nylon filament that is blended with chopped carbon fibers to create a durable/ shatter-resistant filament that is perfect for tough/ functional 3D printed parts.