Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nystatin, 85+%

Catalog No. AC455500050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455500050 5 g
AC455500250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC455500050 Supplier Thermo Scientific Chemicals Supplier No. 455500050
  • $92.00
    $76.65 / Each of 1

    Save $15.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1400-61-9 | C47H75NO17 | 926.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1400-61-9
Molecular Formula C47H75NO17
Molecular Weight (g/mol) 926.107
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
PubChem CID 133640190
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O

Specifications

Type Nystatin
Melting Point >160.0°C
Infrared Spectrum Authentic
Water 6% max.
Quantity 5 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in dmf and dimethylsulfoxide, slightly soluble in methanol, practically insoluble in alcohol
Formula Weight 926.1
Percent Purity ≥85%
Chemical Name or Material Nystatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.