Learn More
Apexbio Technology LLC Nystatin (Fungicidin), 1400-61-9, MFCD27989284, 200 mg
SDP
Catalog No. 501010332
Encompass_Preferred
MF: C47H75NO17, MW: 926.09, Solubility: 30.45 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Nystatin (Fungicidin) is a polyene antifungal antibiotic. Antifungal antibiotic is a pharmaceutical fungicide used to treat and prevent mycoses.
Catalog No. 50-101-0332 Supplier Apexbio Technology LLC Supplier No. B1993200
MF: C47H75NO17, MW: 926.09, Solubility: 30.45 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Nystatin (Fungicidin) is a polyene antifungal antibiotic. Antifungal antibiotic is a pharmaceutical fungicide used to treat and prevent mycoses.