Learn More
Description
- Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests, and defibrination of blood or plasma for serum controls
- Also used for site specific cleavage of recombinant fusion proteins, or in biochemical and medical research applications
- Manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue
- Lyophilized under vacuum in glass vials, or in closed trays as a bulk powder (depending on customer requirements)
- Specific Activity: 90-300 NIH units/mg protein
- Total Protein: > 50%
Specifications
Specifications
| For Use With (Application) | Coagulation Assays, Clotting Factor Tests, Defibrination of Blood/Plasma for Serum Controls, Site Specific Cleavage of Recombinant Fusion Proteins, Biochemical Research, Medical Research |
| Quantity | 500 kU |
| Product Type | Bovine Plasma |
| Specific Reactivity | Specific Activity: 90 to 300 units/mg protein |
| Species | Bovine |
| Content And Storage | Frozen |
| Shipping Condition | Cold Pack |
| Form | Lyophilized |
| Manufacturing Quality | Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.