Learn More
Lonza Walkersville NZ White Hepatocytes Suspens
Catalog No. 5000496360
Encompass
All our suspension cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherant cells
Catalog No. 50-004-96360 Supplier Lonza Walkersville Supplier No. NZCS01
All our suspension cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension They are not suitable for applications requiring adherant cells