Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp NZCYM Broth, 500 Grams
SDP

Catalog No. 50488864
Encompass_Preferred

Enriched medium for cultivating recombinant strains of E.Coli and propagating theta bacteriophage.

Dissolve 22 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Casein Digest Protein: 10 g/L
Yeast Extract: 5 g/L
Casamino Acids: 1 g/L
Magnesium Sulfate: 0.98 g/L
Sodium Chloride: 5 g/L

Catalog No. 50-488-864 Supplier Research Products International Corp Supplier No. N72000500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enriched medium for cultivating recombinant strains of E.Coli and propagating theta bacteriophage.

Dissolve 22 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Casein Digest Protein: 10 g/L
Yeast Extract: 5 g/L
Casamino Acids: 1 g/L
Magnesium Sulfate: 0.98 g/L
Sodium Chloride: 5 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.