Learn More
Bioworld NZCYM Broth, 500 g
Catalog No. 50254443
Encompass_Preferred
NZCYM BrothUsed for the cultivation of various genetic strains of E. coli Formulation recommended for replication of recombinant λ phagepH: 7.0 ±0.2Dehydrated Appearance: light yellow to light brown, homogeneous powderPrepared Appearance: light yellow to yellow Soluble in waterMedia Composition (g/L):Bacto Casein Diges: 10g Bacto Casamino Acids: 1g Bacto Yeast Extract: 5g Sodium Chloride: 5g Magnesium Sulfate: 0.98g
Catalog No. 50-254-443 Supplier Bioworld Supplier No. 306210211
NZCYM BrothUsed for the cultivation of various genetic strains of E. coli Formulation recommended for replication of recombinant λ phagepH: 7.0 ±0.2Dehydrated Appearance: light yellow to light brown, homogeneous powderPrepared Appearance: light yellow to yellow Soluble in waterMedia Composition (g/L):Bacto Casein Diges: 10g Bacto Casamino Acids: 1g Bacto Yeast Extract: 5g Sodium Chloride: 5g Magnesium Sulfate: 0.98g