Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ NZCYM Broth, Capsules

Catalog No. MP113004031 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity: 1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP113004031 Quantity: 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113004031 Supplier MP Biomedicals™ Supplier No. 113004031
Add to Cart
Add to Cart

Culture media used for the growth of bacteria.

  • Used for bacteria cell culture, microbial analysis, gene expression
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Capsules
Media Type Bacterial Media
Product Type Microbiology Media
Quantity 1 kg

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.