Learn More
Sigma Aldrich Fine Chemicals Biosciences NZCYM Broth Powder microbi1KG
Catalog No. 502907792
Encompass
NZCYM is a microbial growth medium used for the cultivation of E. coli and lambda bacteriophage. This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation.
Catalog No. 50-290-7792 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N36431KG
NZCYM is a microbial growth medium used for the cultivation of E. coli and lambda bacteriophage. This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation.