Learn More
Kd Medical Inc NZY BROTH 500ML BOTTLE
SDP
Catalog No. 501018955
Encompass_Preferred
Used for growing microbial/bacterial cells supplemented with nutrients, free amino acids and other components. This product is especially useful for cultivating recombinant strains of E. coli and bacteriophage lambda. One 500ml bottle.
Catalog No. 50-101-8955 Supplier Kd Medical Inc Supplier No. BLE3110
Used for growing microbial/bacterial cells supplemented with nutrients, free amino acids and other components. This product is especially useful for cultivating recombinant strains of E. coli and bacteriophage lambda. One 500ml bottle.