Learn More
Teknova NZYCM Broth, 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387869
Encompass
1.0% NZ Amine, 0.5% Yeast Extract, 0.5% Sodium Chloride, 0.1% Casamino Acids, 10mM Magnesium Sulfate
Catalog No. 50-238-7869 Supplier Teknova Supplier No. N01632PK
1.0% NZ Amine, 0.5% Yeast Extract, 0.5% Sodium Chloride, 0.1% Casamino Acids, 10mM Magnesium Sulfate