Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, 99+%

Catalog No. AAA1836801
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1836801 0.25 g
AAA1836803 1 g
AAA1836806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1836801 Supplier Thermo Scientific Chemicals Supplier No. A1836801
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, >-99%, C7H5ClF5NO, CAS Number-57981-02-9 | CAS: 57981-02-9 | C7H5ClF5NO | 249.57 g/mol

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride is used in the preparation of oximes of steroids having keto group. It is used as a derivatization reagent in the determination of thromboxane B2, prostaglandins, amygdalin and a variety of aldehydes, ketones and acids. It is a versatile reagent utilized for the detection of carbonyl-containing compounds by gas chromatography (GC), gas chromatography mass spectrometry (GC-MS) and other methods. As a derivatization reagent, it is also used for EC/LC analysis of keto-steroids and carbohydrates.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57981-02-9
Molecular Formula C7H5ClF5NO
Molecular Weight (g/mol) 249.57
MDL Number MFCD00012953
InChI Key HVMVKNXIMUCYJA-UHFFFAOYSA-N
Synonym o-2,3,4,5,6-pentafluorobenzyl hydroxylamine hydrochloride, florox reagent, o-perfluorophenyl methyl hydroxylamine hydrochloride, hydroxylamine, o-pentafluorophenyl methyl-, hydrochloride, o-pentafluorobenzyl hydroxylamine hydrochloride, o-2,3,4,5,6-pentafluorophenyl methyl hydroxylamine hydrochloride, oxy 2,3,4,5,6-pentafluorophenyl methyl amine, chloride, pfbha.hcl, acmc-1b0zv
PubChem CID 122307
IUPAC Name O-[(2,3,4,5,6-pentafluorophenyl)methyl]hydroxylamine;hydrochloride
SMILES [H+].[Cl-].NOCC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point ∼215°C (sublimation)
Quantity 0.25 g
Beilstein 4031190
Solubility Information Soluble in water.
Formula Weight 249.57
Percent Purity ≥99%
Chemical Name or Material O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 261-057-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.