Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-[2-(6-Oxocaproylamino)ethyl]-O-methylpolyethylene glycol 2,000 , MilliporeSigma™ Supelco™

Catalog No. 111012678 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2678 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2678 Supplier MilliporeSigma Supelco Supplier No. 54369250MGF

missing translation for 'validMainframeMsg'

Specifications

CAS 1173289-16-1
Packaging Poly Bottle
MDL Number MFCD03792497
Quantity 250 mg
Synonym O-Methyl-O'-[2-(6-oxocaproylamino)ethyl]polyethylene glycol 2'000; PEG-aldehyde 2'000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.