Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-(2-Aminoethyl)-O'-(2-Azidoethyl)Pentaethylene Glycol

Catalog No. 76172500MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-172-500MGF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 76-172-500MGF Supplier Sigma Aldrich Supplier No. 76172500MGF
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-(2-Aminoethyl)-O'sa-(2-azidoethyl)pentaethylene glycol has been used in the synthesis of Alexa568-azide, a fluorescent azide that can undergo click staining reaction with 5-ethynyluridine-labeled cellular RNA.

Specifications

CAS 90852-19-0
Melting Point 123°C to 127°C
Molecular Formula C7H6BFO3
Linear Formula C7H6BFO3
MDL Number MFCD10697421
Quantity 1 g
Synonym Azido-PEG-amine (n=6)
Molecular Weight (g/mol) 167.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.