Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-(2-Aminoethyl)-O'-[2-(Boc-Amino)Ethyl]Hexaethylene Glycol

Catalog No. 70023500MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-023-500MGF 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-023-500MGF Supplier Sigma Aldrich Supplier No. 70023500MGF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Our highly purified mono-protected homobifunctional polyethylene glycols display oligomer purities of more than 90%. This extremely narrow distribution of oligomers ensures homogeneous products in many applications such as conjugation reactions or introduction of solubilizing units or spacers.

Specifications

CAS 151606-29-0
Boiling Point 211°C
Molecular Formula C19H18N2O5S
Linear Formula C19H18N2O5S
MDL Number MFCD03452711
Quantity 5 mg
Molecular Weight (g/mol) 386.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.