Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich O-(2-Aminoethyl)-O'-[2-(Boc-Amino)Ethyl]Octaethylene Glycol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Our highly purified mono-protected homobifunctional polyethylene glycols display oligomer purities of more than 90%. This extremely narrow distribution of oligomers ensures homogeneous products in many applications such as conjugation reactions or introduction of solubilizing units or spacers.
Specifications
Specifications
| CAS | 890091-42-6 |
| Molecular Formula | C29H60N2O13 |
| Linear Formula | C29H60N2O13 |
| Quantity | 500 mg |
| MDL Number | MFCD03453245 |
| Synonym | Boc-PEG-amine (n=11) |
| Molecular Weight (g/mol) | 644.79 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?