Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-(2-Aminoethyl)Polyethylene Glycol 3,000

Catalog No. 07969250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-969-250MG 250 mg
07-969-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-969-250MG Supplier Sigma Aldrich Supplier No. 07969250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-(2-Aminoethyl)polyethylene glycol is a heterobifunctional polyethylene glycol (hydroxyl-PEG-amine) useful in conjugation and crosslinking reactions.O-(2-Aminoethyl)polyethylene glycol has been used to synthesize PEG-poly(D,L-lactide-co-glycolide) (PEG-PLGA) conjugate for use in the synthesis of PLGA-based nanoparticles.

Specifications

CAS 32130-27-1
Linear Formula NH2(CH2CH2O)nH
MDL Number MFCD00286223
Quantity 250 mg
Synonym Aminopolyethylene glycol
Molecular Weight (g/mol) 3000 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.