Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(2-Azido-4,6-O-benzylidene-2-deoxy-α-D-galactopyranosyl)-N-Fmoc-L-serine tert-Butyl Ester 92.0+%, TCI America™
SDP

Catalog No. A1833100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1833100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1833100MG Supplier TCI America Supplier No. A1833100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 878483-02-4
Molecular Formula C35H38N4O9
Molecular Weight (g/mol) 658.71
MDL Number MFCD11112166
InChI Key FJYOABRVKHDUNV-UHFFFAOYNA-N
Synonym O-(2-Azido-4,6-O-benzylidene-2-deoxy-alpha-D-galactopyranosyl)-N-Fmoc-L-serine tert-Butyl Ester, Fmoc-Ser[GalN3[46Bzd]-alpha]-OtBu
PubChem CID 133554188
IUPAC Name tert-butyl 3-({7-azido-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl}oxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
SMILES CC(C)(C)OC(=O)C(COC1OC2COC(OC2C(O)C1N=[N+]=[N-])C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 100 mg
Formula Weight 658.71
Percent Purity ≥92.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(2-Azido-4,6-O-benzylidene-2-deoxy-alpha-D-galactopyranosyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.