Learn More
Medchemexpress LLC O-3M3FBS | 313981-55-4 | MFCD00784362 | >98.0% | 343.36 g/mol | C16H16F3NO2S | 5 MG
SDP
Catalog No. 5000264171
Encompass_Preferred
o-3M3FBS is a research-grade small molecule used as the inactive (negative) control for m-3M3FBS. It modulates ion channel-related currents and intracellular calcium levels, and is used in studies of phospholipase C-related signaling and calcium channel activity.
  • Serves as the negative control for m-3M3FBS in functional assays.
  • Inhibits inward and outward currents by mechanisms independent of phospholipase C.
  • Can also produce an agonist-like increase in intracellular Ca2+ in some systems.
  • High solubility in DMSO (100 mg/mL), suitable for in vitro dosing.
  • Recommended storage: 4°C, protect from light; in solution: -80°C (6 months) or -20°C (1 month).
  • Supplied at high purity (>98%).
  • Suitable for ion channel and calcium signaling research.
Catalog No. 50-002-64171 Supplier Medchemexpress LLC Supplier No. HY1086085MG
o-3M3FBS is a research-grade small molecule used as the inactive (negative) control for m-3M3FBS. It modulates ion channel-related currents and intracellular calcium levels, and is used in studies of phospholipase C-related signaling and calcium channel activity.
  • Serves as the negative control for m-3M3FBS in functional assays.
  • Inhibits inward and outward currents by mechanisms independent of phospholipase C.
  • Can also produce an agonist-like increase in intracellular Ca2+ in some systems.
  • High solubility in DMSO (100 mg/mL), suitable for in vitro dosing.
  • Recommended storage: 4°C, protect from light; in solution: -80°C (6 months) or -20°C (1 month).
  • Supplied at high purity (>98%).
  • Suitable for ion channel and calcium signaling research.