Learn More
Matrix Scientific O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 148893-10-1, MFCD00274639, 25g
Catalog No. 50297448
Encompass_Preferred
Molecular Formula C10H15F6N6OP, Purity 95%, Molecular Weight 380.23, Melting Point ca 185°
Catalog No. 50-297-448 Supplier Matrix Scientific Supplier No. 06722225G
Molecular Formula C10H15F6N6OP, Purity 95%, Molecular Weight 380.23, Melting Point ca 185°