Learn More
Medchemexpress LLC O-(7-azabenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate | 148893-10-1 | MFCD00274639 | 99.9% | 500g
SDP
Catalog No. 5000266245
Encompass_Preferred
HATU (1-[Bis(dimethylamino)methylene]-1H-1 2 3-triazolo[4 5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used in peptide coupling chemistry to generate an active ester from a carboxylic acid HATU can be used along with H nig s base (N N-diisopropylethylamine DIPEA) to form amide bonds[1]
Catalog No. 50-002-66245 Supplier Medchemexpress LLC Supplier No. HYY1703500G
HATU (1-[Bis(dimethylamino)methylene]-1H-1 2 3-triazolo[4 5-b]pyridinium 3-oxid hexafluorophosphate) is a reagent used in peptide coupling chemistry to generate an active ester from a carboxylic acid HATU can be used along with H nig s base (N N-diisopropylethylamine DIPEA) to form amide bonds[1]