Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Acetanisidide 98.0+%, TCI America™
SDP

Catalog No. A001825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A001825G Supplier TCI America Supplier No. A001825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 93-26-5
Melting Point 87°C
Color White
Boiling Point 305°C
Molecular Formula C9H11NO2
Quantity 25 g
MDL Number MFCD00026117
InChI Key FGOFNVXHDGQVBG-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1NC(C)=O
IUPAC Name N-(2-methoxyphenyl)acetamide
Molecular Weight (g/mol) 165.19
PubChem CID 7135
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material o-Acetanisidide
Show More Show Less
EINECSNumber (3)-0690
RTECSNumber AE8280000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.